missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford MTT
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Water soluble salt of Tetrazolium.
Water soluble salt of Tetrazolium. Yields a yellowish solution when prepared in media or salt solutions lacking phenol red. By cleavage of the tetrazolium ring by dehydrogenase enzymes, dissolved MTT is converted into insoluble purple formazan.
Specifications
Specifications
| CAS | 298-93-1 |
| Molecular Formula | C18H16N5SBr |
| Quantity | 1 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction