missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTSS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112617
This item is not returnable.
View return policy
Description
MTSS1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Metastasis suppressor protein 1 (MTSS1) [MIM, missing in metastasis], was first discovered as missing transcripts from metastatic bladder and prostate cancer cells. MTSS1 is a 755 amino acid protein, having multidomain and scaffolding function. It has a proline-rich/SH3 binding region and a serine-rich region with EGF or PDGF/Src consensus sites. It contains anactin-binding Wiskott-Aldrich syndrome protein homology-2 motif (WH2) at the C-terminus while the N-terminal IMD domain (IRSp53-MIM domain) interact with membranes and bundle actin filament. The WH2 domain binds to actin monomers and plays an important role in actin cytoskeleton reorganization. Recently, MIM was found to interact with cortactin via SH3 domains. In addition, MTSS1 enhanced cortactin- and Arp2/3-dependent actin polymerization. It is thus involved in cell motility and morphogenesis and also interacts with sonic hedgehog/Gli signaling in epidermal cells. MTSS1 acts as an important regulator of cell growth and development and is known to express in many tissues, including spleen, thymus, prostate, testis, uterus, colon, and peripheral blood.
Specifications
| MTSS1 | |
| Polyclonal | |
| Unconjugated | |
| MTSS1 | |
| 2310003N14Rik; actin monomer-binding protein; BC024131; D130001D01Rik; hypothetical protein; KIAA0429; metastasis suppressor 1; metastasis suppressor protein 1; Metastasis suppressor YGL-1; Mim; MIMA; MIMB; missing in metastasis protein; mKIAA0429; MTSS I-BAR domain containing 1; MTSS1; MTSS1, I-BAR domain containing; Protein MTSS 1; protein MTSS 1; metastasis suppressor protein 1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 9788 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.84 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O43312 | |
| MTSS1 | |
| Recombinant Human Metastasis suppressor protein 1 protein (616-730AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction