missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTSS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00067-100ul
This item is not returnable.
View return policy
Description
MTSS1 Polyclonal antibody specifically detects MTSS1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| MTSS1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| FLJ44694, KIAA0429Missing in metastasis protein, metastasis suppressor 1, metastasis suppressor protein 1, Metastasis suppressor YGL-1, MIMA, MIMB, MIMDKFZp781P2223, MTSS1 | |
| Produced in rabbits immunized with purified, recombinant Human MTSS1 (Accession#: EAW92073.1; Met1-Ser250) | |
| 100 μg | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 9788 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction