missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mtRNA polymerase Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£165.00 - £365.00
Specifications
| Antigen | mtRNA polymerase |
|---|---|
| Concentration | 1 mg/ml |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18001564
|
Novus Biologicals
NBP3-18334-0.025MG |
0.025 mg |
£165.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18300326
|
Novus Biologicals
NBP3-18334-0.1MG |
0.1 mg |
£365.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mtRNA polymerase Polyclonal antibody specifically detects mtRNA polymerase in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
| mtRNA polymerase | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| APOLMTEC 2.7.7.6, DNA-directed RNA polymerase, mitochondrial, h-mtRPOL, MTRNAP, MTRPOL, polymerase (RNA) mitochondrial (DNA directed) | |
| Antibody was raised against a peptide corresponding to 11 amino acids near the carboxyl terminus of human mtRNA polymerase. The immunogen is located within the last 50 amino acids of mtRNA polymerase. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS | |
| 5442 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title