missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTPG, Tocris Bioscience™
Group II/group III mGlu antagonist. More selective for group II than group III
Chemical Identifiers
CAS | 169209-66-9 |
---|---|
Molecular Formula | C10H11N5O2 |
Molecular Weight (g/mol) | 233.231 |
InChI Key | PUQSGMIEFYCTSW-UHFFFAOYSA-N |
Synonym | mtpg, 2-amino-2-4-tetrazol-1-yl phenyl propanoic acid, 2-amino-2-4-1,2,3,4-tetrazol-1-yl phenyl propanoic acid, +/--alpha-methyl-4-tetrazolylphenyl glycine, inverted exclamation marka-, a-methyl-4-tetrazolylphenyl glycine |
PubChem CID | 5311456 |
IUPAC Name | 2-amino-2-[4-(tetrazol-1-yl)phenyl]propanoic acid |
SMILES | CC(C1=CC=C(C=C1)N2C=NN=N2)(C(=O)O)N |
Description
Group II/group III metabotropic glutamate receptor antagonist, showing selectivity for group II in electrophysiological studies.Chemical Identifiers
169209-66-9 | |
233.231 | |
mtpg, 2-amino-2-4-tetrazol-1-yl phenyl propanoic acid, 2-amino-2-4-1,2,3,4-tetrazol-1-yl phenyl propanoic acid, +/--alpha-methyl-4-tetrazolylphenyl glycine, inverted exclamation marka-, a-methyl-4-tetrazolylphenyl glycine | |
2-amino-2-[4-(tetrazol-1-yl)phenyl]propanoic acid |
C10H11N5O2 | |
PUQSGMIEFYCTSW-UHFFFAOYSA-N | |
5311456 | |
CC(C1=CC=C(C=C1)N2C=NN=N2)(C(=O)O)N |
Specifications
169209-66-9 | |
mtpg, 2-amino-2-4-tetrazol-1-yl phenyl propanoic acid, 2-amino-2-4-1,2,3,4-tetrazol-1-yl phenyl propanoic acid, +/--alpha-methyl-4-tetrazolylphenyl glycine, inverted exclamation marka-, a-methyl-4-tetrazolylphenyl glycine | |
CC(C1=CC=C(C=C1)N2C=NN=N2)(C(=O)O)N | |
233.231 | |
233.23 | |
MTPG |
C10H11N5O2 | |
PUQSGMIEFYCTSW-UHFFFAOYSA-N | |
2-amino-2-[4-(tetrazol-1-yl)phenyl]propanoic acid | |
5311456 | |
>98% |
Safety and Handling
Recommended Storage : Store at Room Temperature