missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mTOR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20657-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to MTOR.
Mammalian target of rapamycin (mTOR) is a serine/threonine kinase that plays a key role in cell growth, cell proliferation, and protein synthesis. mTOR mediates phosphoinositide 3-kinase and Akt/PKB signaling, resulting in phosphorylation of 4EBP1, and initiation of mRNA translation. A second pathway involves regulation of ribosomal S6 kinase, which affects ribosome biogenesis and translation elongation. Mutations affecting the gene results in Smith-Kingsmore syndrome.Specifications
| mTOR | |
| Polyclonal | |
| Unconjugated | |
| MTOR | |
| FRAP, FRAP1, FRAP2, Mammalian target of rapamycin, MTOR, RAFT1, Rapamycin target protein 1, RAPT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2475, 56717, 56718 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P42345, P42346, Q9JLN9 | |
| Mtor | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction