missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mTOR Monoclonal Antibody (F11), eBioscience™, Invitrogen™
GREENER_CHOICE

Product Code. 15257317
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15257317 100 μg 100µg
15247317 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15257317 Supplier Invitrogen™ Supplier No. 14219082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: This F11 monoclonal antibody reacts with human, mouse, and rat mammalian target of rapamycin (mTOR). This 289-kDa serine/threonine kinase is activated via phosphorylation at Ser2448 by the PI3K/Akt signaling pathway in response to growth factors, energy, nutrients, and stress. mTOR is highly conserved amongst eukaryotes and constitutively expressed in immune (e.g. T, B, NK, dendritic cells, mast cells, and neutrophils) and non-immune tissues (e.g. heart). mTOR exists as a member of two distinct complexes, mTORC1 and mTORC2, which have differing sensitivities to rapamycin and phosphorylation targets. Rapamycin inhibits mTOR activity by binding FKBP12, which causes dissociation of mTORC1 and mTORC2. This molecule plays a critical role in cell metabolism, survival, dendritic cell maturation and activation, and protein synthesis. Deregulation of mTOR has been implicated in a variety of cancers. Studies have also suggested a role for mTOR in Foxp3 expression and regulatory T cell differentiation. Applications Reported: This F11 antibody has been reported for use in immunoprecipitation, immunoblotting (WB), and immunohistology staining of paraffin embedded tissue sections. Applications Tested: This F11 antibody has been tested by western blot analysis of reduced cell lysates prepared from Jurkat cells. This can be used at 1-5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mTOR
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone F11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene MTOR
Gene Accession No. P42345, Q9JLN9
Gene Alias 2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS
Gene Symbols MTOR
Host Species Mouse
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2475, 56717
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.