Learn More
Invitrogen™ mTOR Monoclonal Antibody (F11), eBioscience™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 14-2190-37
Description
Description: This F11 monoclonal antibody reacts with human, mouse, and rat mammalian target of rapamycin (mTOR). This 289-kDa serine/threonine kinase is activated via phosphorylation at Ser2448 by the PI3K/Akt signaling pathway in response to growth factors, energy, nutrients, and stress. mTOR is highly conserved amongst eukaryotes and constitutively expressed in immune (e.g. T, B, NK, dendritic cells, mast cells, and neutrophils) and non-immune tissues (e.g. heart). mTOR exists as a member of two distinct complexes, mTORC1 and mTORC2, which have differing sensitivities to rapamycin and phosphorylation targets. Rapamycin inhibits mTOR activity by binding FKBP12, which causes dissociation of mTORC1 and mTORC2. This molecule plays a critical role in cell metabolism, survival, dendritic cell maturation and activation, and protein synthesis. Deregulation of mTOR has been implicated in a variety of cancers. Studies have also suggested a role for mTOR in Foxp3 expression and regulatory T cell differentiation. Applications Reported: This F11 antibody has been reported for use in immunoprecipitation, immunoblotting (WB), and immunohistology staining of paraffin embedded tissue sections. Applications Tested: This F11 antibody has been tested by western blot analysis of reduced cell lysates prepared from Jurkat cells. This can be used at 1-5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%...
FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.
Specifications
| mTOR | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P42345, Q9JLN9 | |
| MTOR | |
| Affinity Chromatography | |
| RUO | |
| 2475, 56717 | |
| 4°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| F11 | |
| Unconjugated | |
| MTOR | |
| 2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS | |
| Mouse | |
| 2 mg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.