missing translation for 'onlineSavingsMsg'
Learn More

mTOR inhibitor-3, MedChemExpress

Product Code. 16477508
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477508 10 mg 10mg
16469987 5 mg 5mg
16403537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16477508 Supplier Medchem Express Supplier No. HY18353
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress mTOR inhibitor-3 is a remarkably selective mTOR inhibitor with a Ki of 1.5 nM. mTOR inhibitor-3 suppresses mTORC1 and mTORC2 in cellular and in vivo pharmacokinetic (PK)/pharmacodynamic (PD) experiments.

mTOR inhibitor-3 is a remarkably selective mTOR inhibitor with a Ki of 1.5 nM. mTOR inhibitor-3 suppresses mTORC1 and mTORC2 in cellular and in vivo pharmacokinetic (PK)/pharmacodynamic (PD) experiments.

Specifications

Chemical Name or Material mTOR inhibitor-3
CAS 1207358-59-5
Color White
Purity Grade Notes Research
Molecular Formula C25H30N8O2
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (105.36 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C2=NC(N3[C@@H](C)COCC3)=C4C(CN(C5=NC=CC=N5)CC4)=N2)C=C1)NCC
Molecular Weight (g/mol) 474.56
Formula Weight 474.56
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.