missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mTOR Human ProcartaPlex™ Simplex Kit

Product Code. 18476638
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18476638 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18476638 Supplier Invitrogen™ Supplier No. EPX01097005901
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The ProcartaPlex Human mTOR Simplex Kit measures the level of total mTOR in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.

The ProcartaPlex Human mTOR Simplex Kit measures the level of total mTOR in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

For preparation of cell lysates, we recommend use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-999999-900) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease and Phosphatase Inhibitor Single–Use Cocktail (100X) (Cat. No. 78440 or 78442). These buffers are not included with the panel. Depending on your cell system, other buffers may be suitable as well.

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum or 50 μL of cell culture supernatant and just four hours to obtain analyzed results.

  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatant, cell lysate, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, xMAP INTELLIFLEX, xMAP INTELLIFLEX DR-SE, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

In addition, the xMAP INTELLIFLEX DR-SE System offers dual-reporter capability, allowing the measurement of a second parameter of the analyte, and features side eject functionalities for added flexibility and automation. Together with ProcartaPlex signaling assays, the xMAP INTELLIFLEX DR-SE instrument enables the quantitation and multiplexing of phosphorylated and total proteins in the same well, on the same Luminex bead, and in the same assay run.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Simplex Kit
Quantity 96 Tests
Species Human
Combinability Combinable
Bead Type Magnetic
Assay Sensitivity As determined for Lot 1: 0.74 U/mL
Assay Range As determined for Lot 1: 2.44 to 10,000 U/mL
Format Simplex Kit
For Use With (Equipment) Luminex™ Instruments
Gene mTOR
Gene Alias MTOR, FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS, mechanistic target of rapamycin, mechanistic target of rapamycin kinase
Gene ID (Entrez) 2475
Gene Symbol mTOR
Interassay CV As determined for Lot 1: 7.63%
Intraassay CV As determined for Lot 1: 8.71%
Product Line ProcartaPlex
Sample Type Cell lysates
Sample Volume Cell lysates (prediluted): 25 μL
UniProt ID P42345
Shipping Condition Wet Ice
Research Area Signaling
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.