missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTO1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-87857
This item is not returnable.
View return policy
Description
MTO1 Polyclonal specifically detects MTO1 in Human samples. It is validated for Western Blot.
Specifications
| MTO1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 25821 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| EC 1.1.1.40, EC 6.3.5, homolog of yeast Mto1, mitochondrial MTO1-3, mitochondrial translation optimization 1 homolog (S. cerevisiae), protein MTO1 homolog, mitochondrial | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of human MTO1 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction