missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR2 Antibody (OTI2D2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-03384
This item is not returnable.
View return policy
Description
MTMR2 Monoclonal specifically detects MTMR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| MTMR2 | |
| Monoclonal | |
| 0.9 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100 | |
| MTMR2 | |
| Full length human recombinant protein of human MTMR2 (NP_057240) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| OTI2D2 | |
| Unconjugated | |
| CMT4B, CMT4B1, EC 3.1.3, EC 3.1.3.2, EC 3.1.3.48, KIAA1073EC 3.1.3.-, myotubularin related protein 2, myotubularin-related protein 2 | |
| Mouse | |
| 73.2 kDa | |
| 0.1 mL | |
| Cancer, Protein Phosphatase | |
| 8898 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction