missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MTMR15 Polyclonal antibody specifically detects MTMR15 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | MTMR15 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | DKFZp451H236, EC 3.1.21.-, EC 3.1.4.1, FANCD2/FANCI-associated nuclease 1DKFZp686K16147, fanconi anemia associated nuclease 1, fanconi-associated nuclease 1, KIAA1018coiled-coil domain-containing protein MTMR15, MTMR15, myotubularin related protein 15, Myotubularin-related protein 15 |
| Host Species | Rabbit |
| Immunogen | myotubularin related protein 15. (Uniprot# Q9Y2M0) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?