missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42677
This item is not returnable.
View return policy
Description
MTMR15 Polyclonal specifically detects MTMR15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| MTMR15 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| DKFZp451H236, EC 3.1.21.-, EC 3.1.4.1, FANCD2/FANCI-associated nuclease 1DKFZp686K16147, fanconi anemia associated nuclease 1, fanconi-associated nuclease 1, KIAA1018coiled-coil domain-containing protein MTMR15, MTMR15, myotubularin related protein 15, Myotubularin-related protein 15 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 22909 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| Q9Y2M0 | |
| FAN1 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human myotubularin related protein 15 (NP_055782.3). | |
| 0.1 mL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction