missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR14 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67561-1-IG-150UL
This item is not returnable.
View return policy
Description
The MTMR14 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MTMR14. This antibody recognizes human, mouse, rat antigen. The MTMR14 antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.Specifications
| MTMR14 | |
| Monoclonal | |
| 1800 μg/mL | |
| C3orf29; FLJ22405; FLJ46453; FLJ90311; HJUMPY; hEDTP | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 74 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1A8B2 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 64419 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction