missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Phosphatidylinositide 3-kinase-derived membrane-anchored phosphatidylinositides, such as phosphatidylinositol 3-phosphate (PtdIns(3)P), regulate diverse cellular processes. The protein encoded by this gene functions as an adaptor subunit in a complex with an active PtdIns(3)P 3-phosphatase. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined.
Specifications
Specifications
| Antigen | MTMR12 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.21 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MTMR12 |
| Gene Accession No. | Q5FVM6, Q80TA6, Q9C0I1 |
| Gene Alias | 3 PAP, 3 phosphatase adapter protein, KIAA1682, MTMR12, PIP3AP |
| Gene Symbols | MTMR12 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?