Learn More
Invitrogen™ MTIF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120180
Description
Positive test controls include: HL-60, Mouse heart, Mouse liver, Mouse kidney, Rat brain. The target is usually found in the following locations: Mitochondrion. Immunogen sequence: MAALFLKRLT LQTVKSENSC IRCFGKHILQ KTAPAQLSPI ASAPRLSFLI HAKAFSTAED TQNEGKKTKK NKTAFSNVGR KISQRVIHLF DEKGNDLGNM HRANVIRLMD ERDLRLVQRN TSTEPAEYQL MTGLQILQER QRLREMEKAN PKTGPTLRKE LILSSNIGQH DLDTKTKQIQ QWIKKKHLVQ ITIKKGKNVD VSENEMEEIF HQILQTMPGI ATFSSRPQAV QGGKALMCVL RAFSKNEEKA YKETQETQER DTLNKDHGND KESNVLHQ.
This gene encodes a translation initiation factor that is involved in mitochondrial protein synthesis. Polymorphism in this gene is associated with the onset of Parkinson's disease. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 5.
Specifications
| MTIF3 | |
| Polyclonal | |
| Unconjugated | |
| MTIF3 | |
| 2810012L14Rik; AI414549; DC38; IF3(mt); IF-3(Mt); IF3mt; IF-3Mt; mitochondrial translational initiation factor 3; Mtif3; Translation initiation factor IF-3, mitochondrial | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 219402, 684274, 76366 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.66 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9CZD5, Q9H2K0 | |
| MTIF3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-278 of human MTIF3 (NP_690876.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.