missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTIF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558930
This item is not returnable.
View return policy
Description
Immunogen sequence: KHLVQITIKK GKNVDVSENE MEEIFHQILQ TMPGIATFSS RPQAVQGGKA LMCVLRALSK NEEKAYKETQ ETQERDTLNK DHGNDKESNV LHQ Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 61%.
This gene encodes a translation initiation factor that is involved in mitochondrial protein synthesis. Polymorphism in this gene is associated with the onset of Parkinson's disease. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 5.
Specifications
| MTIF3 | |
| Polyclonal | |
| Unconjugated | |
| MTIF3 | |
| 2810012L14Rik; AI414549; DC38; IF3(mt); IF-3(Mt); IF3mt; IF-3Mt; mitochondrial translational initiation factor 3; Mtif3; Translation initiation factor IF-3, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 219402 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2K0 | |
| MTIF3 | |
| Recombinant protein corresponding to Human MTIF3. Recombinant protein control fragment (Product #RP-97936). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction