Learn More
Invitrogen™ MTIF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552242
Description
Immunogen sequence: RKEQIPLKPK EKRERDSNVL SVIIKGDVDG SVEAILNIID TYDASHECEL ELVHFGVGDI SANDVNLAET FDGVIYGFNV NAGNVIQQSA AKKGVKIKLH KIIYRLVEDL QEELSSRLPC AVEEHPVGEA SILATFSVTE GKKKVPVAGC Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 85%.
One of the essential components for the initiation of protein synthesis. Protects formylmethionyl-tRNA from spontaneous hydrolysis and promotes its binding to the 30S ribosomal subunits. Also involved in the hydrolysis of GTP during the formation of the 70S ribosomal complex.
Specifications
| MTIF2 | |
| Polyclonal | |
| Unconjugated | |
| MTIF2 | |
| IF2(mt); IF-2(Mt); IF-2mt; mitochondrial translational initiation factor 2; MTIF2; Translation initiation factor IF-2, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4528 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P46199 | |
| MTIF2 | |
| Recombinant protein corresponding to Human MTIF2. Recombinant protein control fragment (Product #RP-101855). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.