missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MTIF2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA548811

Product Code. 15673105

  • £439.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MTIF2 Polyclonal Antibody for Western Blot

One of the essential components for the initiation of protein synthesis. Protects formylmethionyl-tRNA from spontaneous hydrolysis and promotes its binding to the 30S ribosomal subunits. Also involved in the hydrolysis of GTP during the formation of the 70S ribosomal complex.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MTIF2
Polyclonal
Unconjugated
MTIF2
IF2(mt); IF-2(Mt); IF-2mt; mitochondrial translational initiation factor 2; MTIF2; Translation initiation factor IF-2, mitochondrial
Rabbit
Antigen affinity chromatography, Protein A
RUO
4528
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P46199
MTIF2
KLH conjugated synthetic peptide between 138-167 amino acids from the N-terminal region of human MTIF2.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.