missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTHFS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597987
This item is not returnable.
View return policy
Description
MTHFS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Contributes to tetrahydrofolate metabolism. Helps regulate carbon flow through the folate-dependent one-carbon metabolic network that supplies carbon for the biosynthesis of purines, thymidine and amino acids.
Specifications
| MTHFS | |
| Polyclonal | |
| Unconjugated | |
| Mthfs | |
| 1110034I12Rik; 2310020H23Rik; 5, 10-methenyltetrahydrofolate synthetase; 5,10-methenyl-tetrahydrofolate synthetase; 5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase); 5-formyltetrahydrofolate cyclo-ligase; AI119695; FLJ30410; HsT19268; methenyl-THF synthetase; Mthfs | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10588 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P49914 | |
| Mthfs | |
| Recombinant Human 5-formyltetrahydrofolate cyclo-ligase protein (2-203AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction