missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTHFD2L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £443.00
Specifications
| Antigen | MTHFD2L |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:20 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18463211
|
Novus Biologicals
NBP1-82663-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18257625
|
Novus Biologicals
NBP1-82663 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTHFD2L Polyclonal specifically detects MTHFD2L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MTHFD2L | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 441024 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PASHTYVRNKIRAASAVGICSELILKPKDVSQEELLDVTDQLNMDPRVSGILVQLPLPDHVDERTI | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:20 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| FLJ13105, methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like, MGC45532, MGC72244, NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein, probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2 | |
| MTHFD2L | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title