missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTHFD2 Monoclonal Antibody (K70015_7E12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556277
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: TIMKPASISE EELLNLINKL NNDDNVDGLL VQLPLPEHID ERRICNAVSP DKDVDGFHVI NVGRMCLDQY SMLPATPWGV WEIIKRTGIP TLGKNVVVAG RSKNVGMPIA MLLHTDGAHE RPGGDATVTI SHRYTPKEQL KKHTILADIV ISAAGIPNLI TADMIKEGAA VIDVGINRVH DPVTAKPKLV GDVDFEGVRQ
This gene encodes a nuclear-encoded mitochondrial bifunctional enzyme with methylenetetrahydrofolate dehydrogenase and methenyltetrahydrofolate cyclohydrolase activities. The enzyme functions as a homodimer and is unique in its absolute requirement for magnesium and inorganic phosphate. Formation of the enzyme-magnesium complex allows binding of NAD. Alternative splicing results in two different transcripts, one protein-coding and the other not protein-coding. This gene has a pseudogene on chromosome 7.
Specifications
| MTHFD2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P13995 | |
| MTHFD2 | |
| Recombinant human MTHFD2 (immunogen range is 101-300). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K70015_7E12 | |
| Unconjugated | |
| MTHFD2 | |
| AW558851; Bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase, mitochondrial; Methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NAD+ dependent), methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase 2; Mthfd2; NAD-dependent methylene tetrahydrofolate dehydrogenase cyclohydrolase; NAD-dependent methylenetetrahydrofolate dehydrogenase; NMDMC | |
| Mouse | |
| Protein A | |
| RUO | |
| 10797 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction