missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTHFD2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33374-100ul
This item is not returnable.
View return policy
Description
MTHFD2 Monoclonal antibody specifically detects MTHFD2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| MTHFD2 | |
| Monoclonal | |
| Western Blot 1:2000 - 1:8000, ELISA Recommended starting concentration is 1 μg/mL | |
| bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase, methenyltetrahydrofolate cyclohydrolase, methylene tetrahydrofolate dehydrogenase (NAD+ dependent), methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, mitochondrial, NAD-dependent methylene tetrahydrofolate dehydrogenase cyclohydrolase, NMDMC | |
| Recombinant funsion protein containing a sequence corresponding to amino acids 40-250 of human MTHFD2 (NP_006627.2).,, Sequence:, ISGRKLAQQIKQEVRQEVEEWVASGNKRPHLSVILVGENPASHSYVLNKTRAAAVVGINSETIMKPASISEEELLNLINKLNNDDNVDGLLVQLPLPEHIDERRICNAVSPDKDVDGFHVINVGRMCLDQYSMLPATPWGVWEIIKRTGIPTLGKNVVVAGRSKNVGMPIAMLLHTDGAHERPGGDATVTISHRYTPKEQLKKHTILADIV | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10797 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction