missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTHFD1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97940IR
This item is not returnable.
View return policy
Description
MTHFD1 Polyclonal specifically detects MTHFD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| MTHFD1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| C1-THF synthase, EC 6.3.4.3, methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthetase5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methylenetetrahydrofolatecyclohydrolase, 10-formyltetrahydrofolate synthetase, methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1, MTHFCC-1-tetrahydrofolate synthase, cytoplasmic, MTHFDcytoplasmic C-1-tetrahydrofolate synthase | |
| Produced in rabbits immunized with E. coli-derived Human MTHFD1 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 4522 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction