missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102780
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NUDT1.
Oxygen radicals damage chromosomal DNA causing cell death and inducing mutations. Among the various classes of DNA damage caused by oxygen radicals, an oxidized form of guanine base (8-oxoguanine) appears to be important as it can pair with cytosine and adenine and G:C to T:A transversion mutation occurs. A significant amount of 8-OxoG is formed in the chromosomal DNA of mammalian cells, with most damaged nucleotides excised from the DNA and excreted in the urine. Along with 8-oxoG being present in oxidatively damaged DNA, 8-oxo-deoxyguanosine (8-oxo-dGTP) is formed in the nucleotide pool during normal cellular metabolism and following oxidative stress. The 8-oxo-dGTP nucleotide can be incorporated in DNA during polymerization and can result in a mispairing unless repaired. MTH converts 8-oxo-dGTP in the nucleotide pool to the monophosphate and prevents the misincorporation of 8-oxo-dGTP into DNA (Figure courtesy of Dr. Mark Kelley). MTH also recognizes 8-oxo-rGTP, which could incorporate into RNA during gene transcription leading to missense or nonsense protein production.
Specifications
| MTH1 | |
| Polyclonal | |
| Unconjugated | |
| NUDT1 | |
| 2-hydroxy-dATP diphosphatase; 7,8-dihydro-8-oxoguanine triphosphatase; 8-oxo-7,8-dihydro-2'-dGTPase; 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase; 8-oxo-7,8-dihydroguanosine triphosphatase; 8-oxo-dGTPase; Mth1; mutT human homolog (8-oxo-dGTPase); mutT human homolog 1; nucleoside diphosphate-linked moiety X motif 1; nucleoside diphosphate-linked moiety X-type motif 1; nudix (nucleoside diphosphate linked moiety X)-type motif 1; nudix hydrolase 1; nudix motif 1; nudix-type motif 1; NUDT1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 117260, 17766, 4521 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P36639, P53368, P53369 | |
| NUDT1 | |
| A synthesized peptide derived from human NUDT1(Accession P36639), corresponding to amino acid residues G62-G83. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction