missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | MTH1 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18230591
|
Novus Biologicals
NBP2-54664 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662338
|
Novus Biologicals
NBP2-54664-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTH1 Polyclonal specifically detects MTH1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MTH1 | |
| Polyclonal | |
| Rabbit | |
| Base Excision Repair, Cancer, Chromatin Research, DNA Repair, Modulation of DNA Pools, Stem Cell Markers | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 4521 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:RWNGFGGKVQEGETIEDGARRELQEESGLTVDALHKVGQIVFEFVGEPELMDVHVFCTDSIQGTPVESDEMRPCWFQLDQIPFKDMWPDDSYWFPLLLQKKKFHGYFKFQGQDTILDYTL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase, EC 3.6.1.-, MTH1Nucleoside diphosphate-linked moiety X motif 1, mutT human homolog 1,8-oxo-dGTPase, nucleoside diphosphate-linked moiety X-type motif 17,8-dihydro-8-oxoguanine triphosphatase, nudix (nucleoside diphosphate linked moiety X)-type motif 1,8-oxo-7,8-dihydroguanosine triphosphatase, Nudix motif 1 | |
| NUDT1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title