missing translation for 'onlineSavingsMsg'
Learn More

MTH1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18404849 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404849 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404849 Supplier Novus Biologicals Supplier No. NBP297131JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MTH1 Polyclonal antibody specifically detects MTH1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTH1
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase, EC 3.6.1.-, MTH1Nucleoside diphosphate-linked moiety X motif 1, mutT human homolog 1,8-oxo-dGTPase, nucleoside diphosphate-linked moiety X-type motif 17,8-dihydro-8-oxoguanine triphosphatase, nudix (nucleoside diphosphate linked moiety X)-type motif 1,8-oxo-7,8-dihydroguanosine triphosphatase, Nudix motif 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human MTH1 (NP_002443.3; Met1-Val156).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, Chromatin Research, DNA Repair, Modulation of DNA Pools, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4521
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.