missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTGR1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97748PCP
This item is not returnable.
View return policy
Description
MTGR1 Polyclonal antibody specifically detects MTGR1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| MTGR1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| core-binding factor, runt domain, alpha subunit 2, translocated to, 2, EHTDKFZp313F2116, ETO homolog on chromosome 20, ETO homologous on chromosome 20, MTG8-like protein, MTG8-related protein 1, MTGR1p85, myeloid translocation gene-related protein 1, Myeloid translocation-related protein 1, protein CBFA2T2, ZMYND3 | |
| Maps to a region between residue 554 and 604 of human Core-Binding Factor, Runt Domain, Alpha Subunit 2, translocated to, 2 using the numbering given in entry NP_005084.1 (GeneID 9139) | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 9139 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion