missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTG16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540816
This item is not returnable.
View return policy
Description
Peptide sequence: QPRSTPPSMP PPPPAASQGA TRPPSFTPHT LMNGSSHSPT AINGAPCTPN Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 100%; Horse: 85%; Human: 100%; Mouse: 100%; Rat: 100%.
This gene encodes a member of the myeloid translocation gene family which interact with DNA-bound transcription factors and recruit a range of corepressors to facilitate transcriptional repression. The t(16;21)(q24;q22) translocation is one of the less common karyotypic abnormalities in acute myeloid leukemia. The translocation produces a chimeric gene made up of the 5'-region of the runt-related transcription factor 1 gene fused to the 3'-region of this gene. This gene is also a putative breast tumor suppressor. Alternative splicing results in transcript variants.
Specifications
| MTG16 | |
| Polyclonal | |
| Unconjugated | |
| CBFA2T3 | |
| A630044F12Rik; AI465270; AW229127; CBFA2/RUNX1 partner transcriptional co-repressor 3; CBFA2/RUNX1 translocation partner 3; CBFA2T3; Cbfa2t3h; core-binding factor, runt domain, alpha subunit 2, translocated to, 3; core-binding factor, runt domain, alpha subunit 2, translocated to, 3 (human); core-binding factor, runt domain, alpha subunit 2; translocated to, 3; eight twenty one protein 2; ETO/MTG8-related protein ETO-2; Eto2; ETO-2; hMTG16; MTG16; MTG16HEL; MTG8-related gene 2; MTG8-related protein 2; MTGR2; myeloid translocation gene 8 and 16b; myeloid translocation gene on chromosome 16 protein; myeloid translocation gene-related protein 2; protein CBFA2T3; Protein ETO-2; RUNX1T3; translocated to, 3; zinc finger MYND domain-containing protein 4; ZMYND4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 863 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75081 | |
| CBFA2T3 | |
| Synthetic peptide directed towards the N-terminal of human CBFA2T3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction