missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTFR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584278
This item is not returnable.
View return policy
Description
Immunogen sequence: GLHQSTSAVD LIKERREKRA NAGKTLVKNN PKKPEMPNML EILKEMNSVK LRSVKRSEQD V Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 75%.
This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.
Specifications
| MTFR1 | |
| Polyclonal | |
| Unconjugated | |
| Mtfr1 | |
| 1300002C08Rik; 1700080D04Rik; 2810026O10Rik; 4930579E05Rik; chondrocyte protein with a poly-proline region; CHPPR; FAM54A2; Kiaa0009; Mitochondrial fission regulator 1; Mtfr1; RGD1304936 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9650 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15390 | |
| Mtfr1 | |
| Recombinant protein corresponding to Human MTFR1. Recombinant protein control fragment (Product #RP-105034). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction