Learn More
MTFR1 Polyclonal Antibody
GREENER_CHOICE
MTFR1 Polyclonal Antibody
Brand:  Invitrogen PA570577
Product Code. 15878983
  • £441.00 / 100µL
Description
MTFR1 Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MTFR1
Polyclonal
Unconjugated
Mtfr1
1300002C08Rik; 1700080D04Rik; 2810026O10Rik; 4930579E05Rik; chondrocyte protein with a poly-proline region; CHPPR; FAM54A2; Kiaa0009; Mitochondrial fission regulator 1; Mtfr1; RGD1304936
Rabbit
Affinity chromatography
RUO
9650
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q15390
Mtfr1
synthetic peptide directed towards the middle region of human MTFR1 (aa 199-248).