missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTFMT Monoclonal Antibody (OTI1C1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525682
This item is not returnable.
View return policy
Description
MTFMT Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this nuclear gene localizes to the mitochondrion, where it catalyzes the formylation of methionyl-tRNA.
Specifications
| MTFMT | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96DP5 | |
| MTFMT | |
| Human recombinant protein fragment corresponding to amino acids 175-389 of MTFMT produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C1 | |
| Unconjugated | |
| MTFMT | |
| 2310020P08Rik; COXPD15; Fmt; FMT1; LOW QUALITY PROTEIN: methionyl-tRNA formyltransferase, mitochondrial; methionyl-tRNA formyltransferase, mitochondrial; mitochondrial methionyl-tRNA formyltransferase; mitochondrial methionyl-tRNA transformylase; MtFMT; RGD1309462 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 123263 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion