missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTF1 Monoclonal Antibody (OTI7D6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526739
This item is not returnable.
View return policy
Description
MTF1 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a transcription factor that induces expression of metallothioneins and other genes involved in metal homeostasis in response to heavy metals such as cadmium, zinc, copper, and silver. The protein is a nucleocytoplasmic shuttling protein that accumulates in the nucleus upon heavy metal exposure and binds to promoters containing a metal-responsive element (MRE).
Specifications
| MTF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14872 | |
| MTF1 | |
| Human recombinant protein fragment corresponding to amino acids 1-272 of MTF1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7D6 | |
| Unconjugated | |
| MTF1 | |
| metal regulatory transcription factor 1; metal response element binding transcription factor 1; metalloregulatory transcription factor; metal-regulatory transcription factor 1; metal-responsive transcription factor 1; MRE-binding transcription factor; MRE-binding transcription factor-1; Mtf1; MTF-1; Thyls; Transcription factor MTF-1; zinc regulatory factor; ZRF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4520 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction