missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTF1 Antibody (OTI2F3), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72796PE
This item is not returnable.
View return policy
Description
MTF1 Monoclonal antibody specifically detects MTF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MTF1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 4520 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2F3 | |
| Western Blot, Immunohistochemistry | |
| metal regulatory transcription factor 1, metal-regulatory transcription factor 1, metal-responsive transcription factor 1, MGC23036, MRE-binding transcription factor, MRE-binding transcription factor-1, MTF-1, Transcription factor MTF-1, zinc regulatory factor, ZRF | |
| Human recombinant protein fragment corresponding to amino acids 1-272 of human MTF1(NP_005946) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction