missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTF1 Antibody (MTF1/2649), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08238CL1
This item is not returnable.
View return policy
Description
MTF1 Monoclonal antibody specifically detects MTF1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MTF1 | |
| Monoclonal | |
| CoraFluor 1 | |
| metal regulatory transcription factor 1, metal-regulatory transcription factor 1, metal-responsive transcription factor 1, MGC23036, MRE-binding transcription factor, MRE-binding transcription factor-1, MTF-1, Transcription factor MTF-1, zinc regulatory factor, ZRF | |
| Recombinant full-length human MTF1 protein (Uniprot: Q14872) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| MTF1/2649 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4520 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction