missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies mTeSR™1, cGMP
Description
- Use this specialized, feeder-free culture medium to achieve more consistent human pluripotent stem cell (hPSC) cultures with homogenous, undifferentiated phenotypes
- Manufactured under relevant cGMPs, mTeSR1 ensures the highest quality and consistency for reproducible results in your fundamental research, as well as for cell therapy and investigational new drug research applications
- This serum-free, complete cell culture medium is made with pre-screened raw materials to ensure batch-to-batch consistency and robust performance in feeder-free hPSC culture
- Use established protocols for applications ranging from derivation to differentiation with this most widely published feeder-free hPSC culture medium, which has been used by leading pluripotent stem cell researchers to successfully maintain thousands of hPSC lines in over 50 countries
Specifications
Specifications
| Cell Type | Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture; Expansion; Maintenance |
| Quantity | 1 L Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?