missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERFD3 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81741IR
This item is not returnable.
View return policy
Description
MTERFD3 Polyclonal antibody specifically detects MTERFD3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MTERFD3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| FLJ14062, Mitochondrial transcription termination factor 2, Mitochondrial transcription termination factor-like protein, MTERF domain containing 3, mTERF domain-containing protein 3, mitochondrial, mTERF2, mTERFL | |
| MTERFD3 antibody was raised against an 18 amino acid peptide near the amino terminus of human MTERFD3 . The immunogen is located within amino acids 20 - 70 of MTERFD3. Amino Acid Squence: TYTTDKQSSKENTRTVEK | |
| 0.1 mL | |
| Endocrinology | |
| 80298 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction