missing translation for 'onlineSavingsMsg'
Learn More

MTERFD2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18361590 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361590 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361590 Supplier Novus Biologicals Supplier No. NBP281966

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MTERFD2 Polyclonal specifically detects MTERFD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTERFD2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ16261, MGC61716, MTERF domain containing 2, mTERF domain-containing protein 2
Gene Symbols MTERFD2
Host Species Rabbit
Immunogen MTERFD2 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MTERFD2 . The immunogen is located within amino acids 260 - 310 of MTERFD2. Amino Acid Squence: LERLGRYQTPDKKGQTQI
Molecular Weight of Antigen 44 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130916
Test Specificity At least three isoforms of MTERFD2 are known to exist. MTERFD2 antibody is predicted to not cross-react with other MTERFD protein family members.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.