missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERFD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23296-1-AP-150UL
This item is not returnable.
View return policy
Description
Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD1 is composed of three leucine zippers that form a three-stranded coiled-coil that binds to DNA. It binds promoter DNA and acts as a mitochondrial transcription regulator. It has been suggested that only the phosphorylated form of MTERFD1 has transcription termination activity. A closely related mTERF family member, MTERFD3, is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription.Specifications
| MTERFD1 | |
| Polyclonal | |
| Unconjugated | |
| MTERF3 | |
| CGI 12, MTERF domain containing 1, MTERFD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51001, 66410 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R3J4, Q96E29 | |
| MTERF3 | |
| MTERFD1 Fusion Protein Ag19847 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction