missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERFD1 Polyclonal Antibody

MTERFD1 Polyclonal Antibody
Brand: Invitrogen PA576433
This item is not returnable.
View return policy
Description
MTERFD1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| MTERFD1 | |
| Polyclonal | |
| Unconjugated | |
| MTERF3 | |
| 2410017I18Rik; Cgi12; CGI-12; Mitochondrial transcription termination factor 3; MTERF domain containing 1; mTERF domain-containing protein 1, mitochondrial; Mterf3; MTERFD1; transcription termination factor 3, mitochondrial | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51001, 66410 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8R3J4, Q96E29 | |
| MTERF3 | |
| Recombinant fusion protein of human MTERFD1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction