missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTERF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536984
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 39 and 45 kDa. Purity is >95% by SDS-PAGE.
Transcription termination factor. Binds to a 28 bp region within the tRNA(Leu(uur)) gene at a position immediately adjacent to and downstream of the 16S rRNA gene; this region comprises a tridecamer sequence critical for directing accurate termination. Binds DNA along the major grove and promotes DNA bending and partial unwinding. Promotes base flipping. Transcription termination activity appears to be polarized with highest specificity for transcripts initiated on the light strand.
Specifications
| MTERF | |
| Polyclonal | |
| Unconjugated | |
| MTERF1 | |
| 4931431L11Rik; 9230106K09Rik; mitochondrial transcription termination factor; mitochondrial transcription termination factor 1; Mitochondrial transcription termination factor 1a; mitochondrial transcription termination factor 1b; mTERF; Mterf1; mTERF1a; mTERF1b; transcription termination factor 1, mitochondrial; Transcription termination factor 1a, mitochondrial; transcription termination factor, mitochondrial | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 545725, 7978, 85261 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8CHZ9, Q99551, Q9EPI8 | |
| MTERF1, Mterf1a | |
| Synthetic peptide corresponding to amino acids 311-356 of human MTERF. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction