missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTCP1NB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | MTCP1NB |
|---|---|
| Concentration | 0.31 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
This gene was identified by involvement in some t(X;14) translocations associated with mature T-cell proliferations. This region has a complex gene structure, with a common promoter and 5′ exon spliced to two different sets of 3′ exons that encode two different proteins. This gene represents the downstream 8 kDa protein that localizes to mitochondria.Specifications
| MTCP1NB | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P56277, Q61908 | |
| 100272147, 105886298 | |
| MTCP1NB Fusion Protein Ag0338 | |
| Primary | |
| -20°C | |
| CMC4 |
| 0.31 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C6.1B, CMC4, Cx9C motif-containing protein 4, MTCP 1 type A, MTCP1, MTCP1NB, p8, p8MTCP1, Protein p8 MTCP 1 | |
| Cmc4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title