missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTCO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564411
This item is not returnable.
View return policy
Description
Immunogen sequence: LIRAELGQPG NLLGNDHIYN VIV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
Cytochrome c oxidase subunit I (CO1 or MTCO1) is 1 of 3 mitochondrial DNA (mtDNA) encoded subunits (MTCO1, MTCO2, MTCO3) of respiratory Complex IV. Complex IV is located within the mitochondrial inner membrane and is the third and final enzyme of the electron transport chain of mitochondrial oxidative phosphorylation. It is composed of 13 polypeptides. Subunits I, II, and III (MTCO1, MTCO2, MTCO3) are encoded by mtDNA while subunits IV, Va, Vb, VIa, VIb, VIc, VIIa, VIIb, VIIc, and VIII are nuclear encoded. The cytochrome c oxidase family of enzymes have 4 redox centers, 2 hemes and 2 copper centers. In mitochondrial Complex IV, the 2 hemes are a and a3 and the 2 coppers are CuA and CuB. The 2 hemes and CuB are bound to subunit I. Acin-Perez et al. (2003) identified a cell line containing single and double missense mutations in the cytochrome c oxidase (COX) subunit I gene of mouse mitochondrial DNA. And they hypothesized that deleterious mutations can arise and become predominant; cultured cells can maintain several mtDNA haplotypes at stable frequencies; the respiratory chain has little spare COX capacity; and that the size of a cavity in the vicinity of val421 in MTCO1I of animal COX may affect the function of the enzyme.
Specifications
| MTCO1 | |
| Polyclonal | |
| Unconjugated | |
| mt-Co1 | |
| Co1; coi; cox1; coxi; Cytochrome c oxidase polypeptide I; Cytochrome c oxidase subunit 1; cytochrome c oxidase subunit I; mitochondrially encoded cytochrome c oxidase I; mtco1; mt-co1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P00395 | |
| mt-Co1 | |
| Recombinant protein corresponding to Human COX1. Recombinant protein control fragment (Product #RP-103931). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction