missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MTCL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30086
This item is not returnable.
View return policy
Description
MTCL1 Polyclonal antibody specifically detects MTCL1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| MTCL1 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23255 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| CCDC165, KIAA0802, SOGA2 | |
| between 926 and 976 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction