missing translation for 'onlineSavingsMsg'
Learn More

MTCH1 Polyclonal Antibody
GREENER_CHOICE

MTCH1 Polyclonal Antibody

Brand:  Thermo Scientific PA553403

Product Code. 15771341

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MTCH1 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MTCH1
Polyclonal
Unconjugated
MTCH1
2310034O17Rik; AI255158; AU018396; C77849; cell proliferation-inducing protein 60; CGI-64; mitochondrial carrier 1; mitochondrial carrier homolog 1; mitochondrial carrier homolog 1 (C. elegans); mitochondrial carrier-like protein 1; Mtch1; PIG60; presenilin-associated protein; PSAP; SLC25A49; solute carrier family 25, member 49; UNQ1871/PRO4314
Rabbit
Antigen Affinity Chromatography
RUO
23787
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NZJ7
MTCH1
Recombinant protein corresponding to Human MTCH1. Recombinant protein control fragment (Product #RP-90966).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.