missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTCH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69285
This item is not returnable.
View return policy
Description
MTCH1 Polyclonal specifically detects MTCH1 in Human samples. It is validated for Western Blot.
Specifications
| MTCH1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| cell proliferation-inducing protein 60, CGI-64, MGC131998, mitochondrial carrier homolog 1, mitochondrial carrier homolog 1 (C. elegans), Presenilin-associated protein, PSAPPIG60 | |
| Rabbit | |
| 40 kDa | |
| 100 μL | |
| Apoptosis, Lipid and Metabolism | |
| 23787 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NZJ7-2 | |
| MTCH1 | |
| Synthetic peptides corresponding to MTCH1(mitochondrial carrier homolog 1 (C. elegans)) The peptide sequence was selected from the C terminal of MTCH1. Peptide sequence NNCGLQAGLPPYSPVFKSWIHCWKYLSVQGQLFRGSSLLFRRVSSGSCFA. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction