missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTBP Polyclonal Antibody

Description
MTBP Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | MTBP |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Mtbp |
| Gene Accession No. | Q96DY7 |
| Gene Alias | AI429604; hdm2; HDMX; hMTBP; MDM2 (mouse double minute 2)-binding protein, 104kD; MDM2 binding protein; Mdm2, transformed 3T3 cell double minute 2, p53 binding protein binding protein, 104kDa; Mdm2, transformed 3T3 cell double minute p53 binding protein; mdm2-binding protein; MDM2BP; MGC5370; MGC71221; mMTBP; Mtbp; RGD1565672 |
| Gene Symbols | Mtbp |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?