missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MTBP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110909

Product Code. 17138833

  • £550.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DAKELLKYFT SDGLPIGDLQ PLPIQKGEKT FVLTPELSPG KLQVLPFEKA SVCHYHGIEY CLDDRKALER DGGFSELQSR LIRYETQTTC TR Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 93%.

MTBP inhibits cell migration in vitro and suppresses the invasive behavior of tumor cells. The protein may play a role in MDM2-dependent TP53/p53 homeostasis in unstressed cells. The protein inhibits autoubiquitination of MDM2, thereby enhancing MDM2 stability. This promotes MDM2-mediated ubiquitination of TP53/p53 and its subsequent degradation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MTBP
Polyclonal
Unconjugated
Mtbp
AI429604; hdm2; HDMX; hMTBP; MDM2 (mouse double minute 2)-binding protein, 104kD; MDM2 binding protein; Mdm2, transformed 3T3 cell double minute 2, p53 binding protein binding protein, 104kDa; Mdm2, transformed 3T3 cell double minute p53 binding protein; mdm2-binding protein; MDM2BP; MGC5370; MGC71221; mMTBP; Mtbp; RGD1565672
Rabbit
Antigen affinity chromatography
RUO
27085
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96DY7
Mtbp
Recombinant Protein corresonding to Human MTBP. Recombinant protein control fragment (Product #RP-93903).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.