missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MTAP Recombinant Rabbit Monoclonal Antibody (K01_2N40)

Product Code. 30829603
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829603 50 μL
30827846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829603 Supplier Invitrogen™ Supplier No. MA565590
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DPEILEGRTE KYVD.

This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage of both adenine and methionine. The encoded enzyme is deficient in many cancers because this gene and the tumor suppressor p16 gene are co-deleted. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length natures remain unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTAP
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2N40
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene MTAP
Gene Accession No. Q13126, Q9CQ65
Gene Alias 1300019I21Rik; 5'-methylthioadenosine phosphorylase; BDMF; c86fus; Cc1-6; DMSFH; DMSMFH; epididymis luminal protein 249; HEL-249; LGMBF; MeSAdo phosphorylase; methylthioadenosine phosphorylase; MSAP; MTA phosphorylase; MTAP; MTAPase; S-methyl-5'-thioadenosine phosphorylase; zgc:66012
Gene Symbols MTAP
Host Species Rabbit
Immunogen A synthetic peptide of human MTAP (aa 22-35).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298227, 4507, 66902
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.